Novus Biologicals
Manufacturer Code:NBP298256FR
Catalog # NB053032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Serpin A7/TBG (NP_808588.3 Met 1-Gln 418). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antitrypsin) member 7 FLJ17820 member 7 Serpin A7 serpin peptidase inhibitor clade A (alpha-1 antiproteinase antitrypsin) T4-binding globulin TBGclade A (alpha-1 antiproteinase thyroxin-binding globulin thyroxine-binding globulin; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 7; Serpin A7; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 7; T4-binding globulin; thyroxin-binding globulin; Thyroxine-binding globulin
Gene Aliases: SERPINA7; TBG
UniProt ID: (Human) P05543
Entrez Gene ID: (Human) 6906
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.