Novus Biologicals
Manufacturer Code:NBP30334920UL
Catalog # NB095453
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-207 of human Serotonin N-acetyltransferase (NP_001079.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA-NAT; AA-NAT aralkylamine N-acetyltransferaseEC 2.3.1.87 arylalkylamine N-acetyltransferase Serotonin acetylase SNATserotonin N-acetyltransferase; Aralkylamine N-acetyltransferase; arylalkylamine N-acetyltransferase; Serotonin acetylase; Serotonin N-acetyltransferase
Gene Aliases: AANAT; DSPS; SNAT
UniProt ID: (Human) Q16613
Entrez Gene ID: (Human) 15
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.