Novus Biologicals
Manufacturer Code:NBP274083R
Catalog # NBP274083R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E3 |
Immunogen | Full length recombinant protein of human Serine racemase (NP_068766) produced in HEK293T cells. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: D-serine ammonia-lyase; D-serine ammonia-lyase D-serine dehydratase EC 4.3.1.17 EC 4.3.1.18 EC 5.1.1.18 ILV1 ISO1 L-serine ammonia-lyase L-serine dehydratase serine racemase; D-serine dehydratase; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase
Gene Aliases: ILV1; ISO1; SRR
UniProt ID: (Human) Q9GZT4
Entrez Gene ID: (Human) 63826
Molecular Function:
deaminase
dehydratase
epimerase/racemase
hydrolase
isomerase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.