Novus Biologicals
Manufacturer Code:NBP293173100UL
Catalog # NB028233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human SRR (NP_068766.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-serine ammonia-lyase; D-serine ammonia-lyase D-serine dehydratase EC 4.3.1.17 EC 4.3.1.18 EC 5.1.1.18 ILV1 ISO1 L-serine ammonia-lyase L-serine dehydratase serine racemase; D-serine dehydratase; L-serine ammonia-lyase; L-serine dehydratase; Serine racemase
Gene Aliases: ILV1; ISO1; SRR
UniProt ID: (Human) Q9GZT4
Entrez Gene ID: (Human) 63826
Molecular Function:
deaminase
dehydratase
epimerase/racemase
hydrolase
isomerase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.