Novus Biologicals
Manufacturer Code:NBP29370720UL
Catalog # NB029278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 423-562 of human SPTLC2 (NP_004854.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.3.1 EC 2.3.1.50 KIAA0526HSN1C LCB 2 LCB2a LCB2NSAN1C Long chain base biosynthesis protein 2 Long chain base biosynthesis protein 2a serine palmitoyltransferase 2 serine palmitoyltransferase long chain base subunit 2 serine palmitoyltransferase subunit II Serine-palmitoyl-CoA transferase 2 SPT 2 SPT2; LCB 2; LCB2a; Long chain base biosynthesis protein 2; Long chain base biosynthesis protein 2a; Serine palmitoyltransferase 2; serine palmitoyltransferase, long chain base subunit 2; serine palmitoyltransferase, subunit II; Serine-palmitoyl-CoA transferase 2; SPT 2
Gene Aliases: hLCB2a; HSN1C; KIAA0526; LCB2; LCB2A; NSAN1C; SPT2; SPTLC2
UniProt ID: (Human) O15270
Entrez Gene ID: (Human) 9517
Molecular Function:
transaminase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.