Invitrogen
Manufacturer Code:PA5113691
Catalog # PIPA5113691
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: LLKCSRLVLA LALILVLESS VQGYPTRRAR YQWVRCNPDS NSANCLEEKG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gp600 hematopoetic proteoglycan core peptide hematopoetic proteoglycan core protein Hematopoietic proteoglycan core protein Mastocytoma proteoglycan core protein P.PG Platelet proteoglycan core protein PPG Prg PRG1 proteoglycan 1 secretory granule proteoglycan protein core for mast cell secretory granule proteoglycan secretory granule secretory granule proteoglycan core peptide Secretory granule proteoglycan core protein Serglycin serglycin proteoglycan Sgc Srgn; hematopoetic proteoglycan core peptide; hematopoetic proteoglycan core protein; Hematopoietic proteoglycan core protein; P.PG; Platelet proteoglycan core protein; proteoglycan 1, secretory granule; proteoglycan protein core for mast cell secretory granule; secretory granule proteoglycan core peptide; Secretory granule proteoglycan core protein; Serglycin; serglycin proteoglycan
Gene Aliases: PPG; PRG; PRG1; SRGN
UniProt ID: (Human) P10124
Entrez Gene ID: (Human) 5552
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.