Invitrogen
Manufacturer Code:PA554755
Catalog # PIPA554755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Septin-7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC10 (cell division cycle 10, S.cerevisiae, homolog); CDC10 protein homolog; cell division cycle 10 homolog; SEPT7 septin 7 CDC10 CDC3 Nbla02942 SEPT7A CDC10 (cell division cycle 10 S. cerevisiae homolog) CDC10 cell division cycle 10 homolog cell division cycle 10; septin 7 variant 4; Septin-7
Gene Aliases: CDC10; CDC3; E430034N22; NBLA02942; SEPT7; SEPT7A; SEPTIN7
UniProt ID: (Human) Q16181, (Mouse) O55131, (Rat) Q9WVC0
Entrez Gene ID: (Human) 989, (Mouse) 235072, (Rat) 64551
Molecular Function:
G-protein
cytoskeletal protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.