Novus Biologicals
Manufacturer Code:NB100330
Catalog # NB100330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | XJ11-4D7 |
Immunogen | Maltose-Binding Protein fusion of C-terminal fragment of human Separase (amino acids 1866-1996). [UniProt# Q14674] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Caspase-like protein ESPL1; Caspase-like protein ESPL1 EC 3.4.22.49 ESP1extra spindle poles like 1 extra spindle pole bodies homolog 1 (S. cerevisiae) extra spindle poles like 1 (S. cerevisiae) Extra spindle poles-like 1 protein FLJ46492 KIAA0165separin Separase; extra spindle pole bodies 1, separase; extra spindle pole bodies homolog 1; extra spindle poles like 1; Extra spindle poles-like 1 protein; Separase; Separin; separin, cysteine protease
Gene Aliases: ESP1; ESPL1; KIAA0165; SEPA
UniProt ID: (Human) Q14674
Entrez Gene ID: (Human) 9700
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.