Invitrogen
Manufacturer Code:PA519419
Catalog # PIPA519419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from residues 1200 - 1300 of Human Separase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Caspase-like protein ESPL1; Cut1/ESP1 related protein; ESP1 FLJ46492 KIAA0165 caspase-like protein ESPL1 extra spindle poles like 1 extra spindle poles-like 1 protein separin; extra spindle pole bodies 1, separase; extra spindle pole bodies homolog 1; extra spindle poles like 1; Extra spindle poles-like 1 protein; Separase; Separin; separin, cysteine protease
Gene Aliases: AL024103; AU045071; Cerp; ESP1; ESPL1; KIAA0165; PRCE; SEPA; SSE
UniProt ID: (Human) Q14674, (Mouse) P60330
Entrez Gene ID: (Human) 9700, (Mouse) 105988
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.