Novus Biologicals
Manufacturer Code:NB10041107
Catalog # NB10041107
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence DEATKKACYSKGKS corresponding to internal region according to NP_003957.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ12815 sema domain seven thrombospondin repeats (type 1 and type 1-like) Sema F SEMAF semaphorin F semaphorin-5A Semaphorin-F semF transmembrane domain (TM) and short cytoplasmic domain (semaphorin) 5A; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; Sema F; semaphorin F; Semaphorin-5A; Semaphorin-F
Gene Aliases: SEMA5A; SEMAF; semF
UniProt ID: (Human) Q13591
Entrez Gene ID: (Human) 9037
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.