R&D Systems
Manufacturer Code:MAB74701SP
Catalog # MAB74701SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 758726 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Semaphorin 4D/CD100 Met22-Arg734 Accession # Q92854 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A8; A8 BB18 CD100 CD100 antigen CD100M-sema-G coll-4 FLJ33485 FLJ34282 FLJ39737 FLJ46484 immunoglobulin domain (Ig) transmembrane domain (TM) and shortcytoplasmic domain 4D MGC169138 MGC169141 M-sema-G sema domain immunoglobulin domain (Ig) transmembrane domain (TM) and shortcytoplasmic domain (semaphorin) 4D SEMA4D SEMAJ semaphorin-4D; BB18; CD100; GR3; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; Semaphorin-4D
Gene Aliases: C9orf164; CD100; coll-4; M-sema-G; SEMA4D; SEMAJ
UniProt ID: (Human) Q92854
Entrez Gene ID: (Human) 10507
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.