R&D Systems
Manufacturer Code:FAB8497V100UG
Catalog # FAB8497V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 663203 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4C Ala21-Phe664 Accession # Q64151 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: FLJ20369 KIAA1739 MGC126382 MGC126383 M-Sema F M-SEMA-F sema domain immunoglobulin domain (Ig) transmembrane domain (TM) and shortcytoplasmic domain (semaphorin) 4C SEMA4C SEMACL1 SEMAF SEMAI semaphorin-4C; M-Sema F; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4C; Semaphorin-4C
Gene Aliases: KIAA1739; M-SEMA-F; SEMA4C; SEMACL1; SEMAF; SEMAI; UNQ5855/PRO34487
UniProt ID: (Human) Q9C0C4
Entrez Gene ID: (Human) 54910
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.