Novus Biologicals
Manufacturer Code:NBP184408
Catalog # NBP184408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:MDLGLLFLRLHKSDAGTYFCQTVEHSFVHTVRKITLEVVEEEKVEDMFNKDDEEDRHHRMPCPAQSSISQGAKPWYKEFLQLIGYSNFQRVEEYCEKVWCTDRKRKKLKMSPSKWKYANPQEKKLRSKPEHYR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (semaphorin) 3E coll-5 KIAA0331M-SEMAH sema domain immunoglobulin domain (Ig) short basic domain secreted SEMAHM-SemaK semaphorin-3E; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E; Semaphorin-3E
Gene Aliases: coll-5; KIAA0331; M-SEMAH; M-SemaK; SEMA3E; SEMAH
UniProt ID: (Human) O15041
Entrez Gene ID: (Human) 9723
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.