R&D Systems
Manufacturer Code:IC1250S100UG
Catalog # IC1250S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 215803 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3A Lys26-Val771 (Arg555Ala Arg552Ala) Accession # Q14563 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: (semaphorin) 3A coll-1 collapsin 1 Hsema-I Hsema-III MGC133243 sema domain immunoglobulin domain (Ig) short basic domain secreted Sema III SEMA1 Sema3A SEMAD SEMAIII SEMAL semaphorin D Semaphorin III semaphorin L semaphorin-3A SemD; collapsin 1; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A; Sema III; semaphorin D; Semaphorin III; semaphorin L; Semaphorin-3A
Gene Aliases: coll-1; COLL1; HH16; Hsema-I; Hsema-III; SEMA1; SEMA3A; SEMAD; SEMAIII; SEMAL; SemD
UniProt ID: (Human) Q14563
Entrez Gene ID: (Human) 10371
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.