Novus Biologicals
Manufacturer Code:NBP184409
Catalog # NBP184409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VPRLKLSYKEMLESNNVITFNGLANSSSYHTFLLDEERSRLYVGAKDHIFSFDLVNIKDFQKIVWPVSYTRRDECKWAGKDILKECANFIKVL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (semaphorin) 3A coll-1 collapsin 1 Hsema-I Hsema-III MGC133243 sema domain immunoglobulin domain (Ig) short basic domain secreted Sema III SEMA1 SEMAD SEMAIII SEMAL semaphorin D Semaphorin III semaphorin L semaphorin-3A SemD; collapsin 1; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A; Sema III; semaphorin D; Semaphorin III; semaphorin L; Semaphorin-3A
Gene Aliases: coll-1; COLL1; HH16; Hsema-I; Hsema-III; SEMA1; SEMA3A; SEMAD; SEMAIII; SEMAL; SemD
UniProt ID: (Human) Q14563
Entrez Gene ID: (Human) 10371
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.