Invitrogen
Manufacturer Code:PA512278
Catalog # PIPA512278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide selected from the N-terminal region of mouse Sema5a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: M-Sema D; sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A; Sema F; semaphorin F; Semaphorin-5A; Semaphorin-5A Semaphorin-F Sema F SEMA5A SEMAF; Semaphorin-F
Gene Aliases: 5930434A13; 9130201M22Rik; AI464145; SEMA5A; SEMAF; semF
UniProt ID: (Human) Q13591
Entrez Gene ID: (Human) 9037, (Mouse) 20356
Molecular Function:
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.