Abnova
Manufacturer Code:MAB6583
Catalog # 89-112-946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SK-3 |
Immunogen | Recombinant protein corresponding to extracellular region of mouse Sema4d. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant Sema4d.
For Research Use Only
Protein Aliases: CD100; coll-4; M-Sema G; Sema J; Semacl2; Semaj; semaphorin H; Semaphorin-4D; Semaphorin-C-like 2; Semaphorin-J; Semcl2
Gene Aliases: CD100; coll-4; Sema4d; Semacl2; Semaj; Semcl2
UniProt ID: (Mouse) O09126
Entrez Gene ID: (Mouse) 20354
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.