Abnova
Manufacturer Code:PAB9009
Catalog # 89-134-089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 756 of mouse Selplg. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Selplg.
For Research Use Only
Protein Aliases: CD162; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand-1; Psgl-1; Psgl1; selectin P ligand; Selp1; Selpl
Gene Aliases: CD162; Psgl-1; Psgl1; Selp1; Selpl
Entrez Gene ID: (Mouse) 20345
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.