Novus Biologicals
Manufacturer Code:NBP2471380.2MG
Catalog # NBP24713802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SC05/COMPO2 |
Immunogen | Partially purified preparation of Secretory Component from human colostrum. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Free Secretory Component (FSC) pIgA-Secretory Component Complex pIgM-Secretory Component Complex Secretory Component (SC) sIgA-Secretory Component Complex; hepatocellular carcinoma associated protein TB6; Hepatocellular carcinoma-associated protein TB6; PIgR; poly-Ig receptor; Polymeric immunoglobulin receptor; Secretory component
Gene Aliases: PIGR
UniProt ID: (Human) P01833
Entrez Gene ID: (Human) 5284
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.