Invitrogen
Manufacturer Code:PA569182
Catalog # PIPA569182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human Secretin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: prepro-secretin; Secretin; secretin SCT
Gene Aliases: SCT; Secr
UniProt ID: (Human) P09683, (Rat) P11384, (Mouse) Q08535
Entrez Gene ID: (Horse) 100630455, (Human) 6343, (Rat) 24769, (Guinea pig) 100733638, (Mouse) 20287
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.