Novus Biologicals
Manufacturer Code:NBP1974920.025MG
Catalog # NB018786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 14G1 |
Immunogen | Recombinant rat rSec8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Exocyst complex component 4; exocyst complex component 4 KIAA1699SEC8L1 MGC27170 REC8 SEC8Exocyst complex component Sec8 SEC8-like 1 SEC8-like 1 (S. cerevisiae) Sec8p; Exocyst complex component Sec8; SEC8-like 1
Gene Aliases: EXOC4; KIAA1699; SEC8; SEC8L1; Sec8p
UniProt ID: (Human) Q96A65
Entrez Gene ID: (Human) 60412
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.