Proteintech
Manufacturer Code:119131AP150UL
Catalog # 50-172-7555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Sec8 Fusion Protein Ag2514 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: augmenter of liver regeneration (ALR) pseudogene; EXOC4 exocyst complex component 4 Exocyst complex component Sec8 KIAA1699 REC8 Sec8 SEC8L1 Sec8p; Exocyst complex component 4; Exocyst complex component Sec8; rSec8; SEC8-like 1; secretory protein SEC8
Gene Aliases: ALR; C78892; EXOC4; KIAA1699; SEC8; SEC8L1; Sec8p
UniProt ID: (Human) Q96A65, (Rat) Q62824, (Mouse) O35382
Entrez Gene ID: (Human) 60412, (Rat) 116654, (Mouse) 20336
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.