Novus Biologicals
Manufacturer Code:NBP293433
Catalog # NB028743
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human SCRIB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein LAP4; Protein scribble homolog; protein scribble homolog CRIB1 SCRB1 SCRIB1 scribbled homolog (Drosophila) Vartul; Scribble; scribbled homolog
Gene Aliases: CRIB1; KIAA0147; LAP4; SCRB1; SCRIB; SCRIB1; VARTUL
UniProt ID: (Human) Q14160
Entrez Gene ID: (Human) 23513
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.