Abnova
Manufacturer Code:PAB28805
Catalog # 89-130-327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Scnn1g C-terminus (amino acids 629-650). |
Conjugate | ATTO 565 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant Rat Scnn1g.
For Research Use Only
Protein Aliases: amiloride sensitive sodium channel gamma1 subunit; Amiloride-sensitive sodium channel subunit gamma; ENaC; Epithelial Na(+) channel subunit gamma; epithelial sodium channel gamma subunit; Gamma-ENaC; Gamma-NaCH; Nonvoltage-gated sodium channel 1 subunit gamma; SCNEG; Sodium channel nonvoltage-gated 1 gamma (epithelial); Sodium channel, nonvoltage-gated 1, gamma (epithelial); sodium channel, nonvoltage-gated, type I, gamma
Gene Aliases: ENaC; Scnn1g
UniProt ID: (Rat) P37091
Entrez Gene ID: (Rat) 24768
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.