Abnova
Manufacturer Code:MAB16462
Catalog # 89-999-419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S395-68 |
Immunogen | Recombinant protein corresponding to full length mouse Scn2b. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2810451E09Rik; AI840361; Gm183; MGC116602; Sodium channel subunit beta-2; sodium channel, voltage-gated, type II, beta polypeptide
Gene Aliases: 2810451E09Rik; AI840361; Gm183; Scn2b
UniProt ID: (Mouse) Q56A07
Entrez Gene ID: (Mouse) 72821
Molecular Function: cell adhesion molecule ion channel sodium channel transporter voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.