EMD Millipore
Manufacturer Code:MABS445
Catalog # MABS445MI
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7B6.1 |
Immunogen | GST-tagged recombinant protein corresponding to the internal fragment of human Sclerostin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | &minus20°C for one year from date of shipment Upon first thaw,and prior to removing the cap,centrifuge the vial and gently mix the solution Aliquot into microcentrifuge tubes and store at &minus20°C Avoid repeated freeze/thaw cycles |
Mouse Monoclonal Antibody
For Research Use Only
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.