Invitrogen
Manufacturer Code:PA5113315
Catalog # PIPA5113315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Sclerostin protein (24-213AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5430411E23Rik CDD DAND6 sclerosteosis Sclerostin Sost SOST1 UNQ2976/PRO7455/PRO7476 VBCH; Sclerostin
Gene Aliases: CDD; DAND6; SOST; SOST1; UNQ2976/PRO7455/PRO7476; VBCH
UniProt ID: (Human) Q9BQB4
Entrez Gene ID: (Human) 50964
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.