Invitrogen
Manufacturer Code:51972980
Catalog # 51-972-980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | NRNSTNX |
Immunogen | C-terminal fragment of the protein (aa-805-1322) |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4°C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 89 HEL-S-89 HSal1 Msal-3 SAL1 Sal-1 sal-3 SALL1 sal-like 1 (Drosophila) Sal-like protein 1 spalt like transcription factor 1 Spalt-like transcription factor 1 TBS Zinc finger protein 794 Zinc finger protein SALL1 Zinc finger protein Spalt-1 zinc finger protein Spalt-3 ZNF794; sal-3; sal-like protein 1; zinc finger protein Spalt-3
Gene Aliases: Msal-3
Entrez Gene ID: (Mouse) 58198
Molecular Function: nucleic acid binding transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.