Novus Biologicals
Manufacturer Code:NBP261869
Catalog # NBP2-61869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G4D8 |
Immunogen | Purified recombinant fragment of human AMY1A (AA: 172-284) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase 1; 14-alpha-D-glucan glucanohydrolase 1 alpha-amylase 1 AMY1 AMY1B AMY1C amylase alpha 1A (salivary) amylase alpha 1A salivary amylase salivary alpha-1A EC 3.2.1.1 glycogenase Salivary alpha-amylase salivary amylase alpha 1A; Alpha-amylase 1A; Alpha-amylase 1B; Alpha-amylase 1C; amylase, salivary, alpha-1A; glycogenase; Salivary alpha-amylase; salivary amylase alpha 1A
Gene Aliases: AMY1; AMY1A; AMY1B; AMY1C
UniProt ID: (Human) P0DUB6
Entrez Gene ID: (Human) 276
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.