Abnova
Manufacturer Code:H00084447M01A
Catalog # 89-107-452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4H4 |
Immunogen | SYVN1 (NP_079434238 a.a. &bsim 318 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant SYVN1.
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase synoviolin; HMG-coA reductase degradation 1 homolog; HRD1; KIAA1810; MGC40372; RING-type E3 ubiquitin transferase synoviolin; Synovial apoptosis inhibitor 1; synovial apoptosis inhibitor 1, synoviolin
Gene Aliases: DER3; HRD1; KIAA1810; SYVN1
UniProt ID: (Human) Q86TM6
Entrez Gene ID: (Human) 84447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.