Novus Biologicals
Manufacturer Code:NBP29322820UL
Catalog # NB028344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 38-170 of human SYT7 (NP_004191.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IPCA-7; IPCA-7synaptotagmin 7 MGC150517 prostate cancer associated protein 7 Prostate cancer-associated protein 7 synaptotagmin VIIPCANAP7 synaptotagmin-7 sytVII SYT-VII; Prostate cancer-associated protein 7; Synaptotagmin VII; Synaptotagmin-7; SytVII
Gene Aliases: IPCA-7; IPCA7; PCANAP7; SYT-VII; SYT7; SYTVII
UniProt ID: (Human) O43581
Entrez Gene ID: (Human) 9066
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.