Invitrogen
Manufacturer Code:PA539488
Catalog # PIPA539488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human SYT11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0080; synaptotagmin 11; synaptotagmin 12; Synaptotagmin XI; Synaptotagmin-11; SytXI
Gene Aliases: 1500004A13Rik; 3632445O20Rik; 5430404N14Rik; 6530420C11Rik; AI851970; KIAA0080; SYT11; SYT12; sytXI
UniProt ID: (Human) Q9BT88, (Mouse) Q9R0N3, (Rat) O08835
Entrez Gene ID: (Human) 23208, (Mouse) 229521, (Rat) 60568
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.