Proteintech
Manufacturer Code:156661AP150UL
Catalog # 50-173-0389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SYNJ2BP Fusion Protein Ag8130 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activin receptor interacting protein 2; activin receptor interacting protein 5; Activin receptor-interacting protein 2; activin receptor-interacting protein 2a; Activin receptor-interacting protein 4; ARIP2 OMP25 synaptojanin 2 binding protein SYNJ2BP; Mitochondrial outer membrane protein 25; outer membrane protein 25; Synaptojanin-2-binding protein; synj2bp
Gene Aliases: AA409442; ActRIP4; ARIP2; ARIP2a; D12Wsu118e; OMP25; SYNJ2BP
UniProt ID: (Human) P57105, (Mouse) Q9D6K5
Entrez Gene ID: (Human) 55333, (Mouse) 24071
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.