Invitrogen
Manufacturer Code:PA524310
Catalog # PIPA524310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 852-880 amino acids from the C-terminal region of human SYNE3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KASH domain-containing protein 3; KASH3; Nesprin-3; NET53 SYNE3 nesprin-3; Nuclear envelope spectrin repeat protein 3; spectrin repeat containing, nuclear envelope family member 3
Gene Aliases: C14orf139; C14orf49; LINC00341; Nesp3; nesprin-3; NET53; SYNE3
UniProt ID: (Human) Q6ZMZ3, (Mouse) Q4FZC9
Entrez Gene ID: (Human) 161176, (Mouse) 212073
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.