Proteintech
Manufacturer Code:252651AP150UL
Catalog # 50-173-4823
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KASH domain-containing protein 2; KASH2; KIAA1011 Nesprin 2 NUA NUANCE Protein NUANCE SYNE 2 SYNE2; nesprin 2; Nesprin-2; Nuclear envelope spectrin repeat protein 2; Nucleus and actin connecting element protein; polytrophin; Protein NUANCE; spectrin repeat containing, nuclear envelope 2; Synaptic nuclear envelope protein 2; synaptic nuclei expressed gene 2; Syne-2
Gene Aliases: EDMD5; KIAA1011; Nesp2; Nesprin-2; NUA; NUANCE; SYNE-2; SYNE2; TROPH
UniProt ID: (Human) Q8WXH0
Entrez Gene ID: (Human) 23224
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.