OriGene
Manufacturer Code:CF811708
Catalog # 50-167-0797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-285 of human SYNE1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 8B ARCA1 C6orf98 CPG2 EDMD4 KASH1 MYNE1 Nesp1 SCAR8 dJ45H2.2; CPG2 full length; Enaptin; KASH domain-containing protein 1; KASH1; Myne-1; Myocyte nuclear envelope protein 1; nesprin 1; Nesprin-1; Nuclear envelope spectrin repeat protein 1; spectrin repeat containing, nuclear envelope 1; Synaptic nuclear envelope protein 1; synaptic nuclei expressed gene 1; Syne-1
Gene Aliases: 8B; ARCA1; C6orf98; CPG2; dJ45H2.2; EDMD4; KIAA0796; KIAA1262; KIAA1756; MYNE1; Nesp1; SCAR8; SYNE1
UniProt ID: (Human) Q8NF91
Entrez Gene ID: (Human) 23345
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.