Novus Biologicals
Manufacturer Code:NBP298081C
Catalog # NB052273
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SYNE1 fragment. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARCA1 C6orf98 CPG2 dJ45H2.2 EC 4.2.1.49 EC 4.3.1.3 EDMD4 Enaptin KIAA0796chromosome 6 open reading frame 98 KIAA1262FLJ41140 KIAA17568B Myne-1 MYNE1DKFZp781J13156 Myocyte nuclear envelope protein 1 nesprin 1 nesprin-1 Nuclear envelope spectrin repeat protein 1 SCAR8FLJ30878 spectrin repeat containing nuclear envelope 1 Synaptic nuclear envelope protein 1 synaptic nuclei expressed gene 1 Syne-1 SYNE-1B; CPG2 full length; Enaptin; KASH domain-containing protein 1; KASH1; Myne-1; Myocyte nuclear envelope protein 1; nesprin 1; Nesprin-1; Nuclear envelope spectrin repeat protein 1; spectrin repeat containing, nuclear envelope 1; Synaptic nuclear envelope protein 1; synaptic nuclei expressed gene 1; Syne-1
Gene Aliases: 8B; ARCA1; C6orf98; CPG2; dJ45H2.2; EDMD4; KIAA0796; KIAA1262; KIAA1756; MYNE1; Nesp1; SCAR8; SYNE1
UniProt ID: (Human) Q8NF91
Entrez Gene ID: (Human) 23345
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.