Abnova
Manufacturer Code:MAB2498
Catalog # 89-111-037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | I8E4 |
Immunogen | Recombinant protein corresponding to human SYNCRIP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant SYNCRIP.
For Research Use Only
Protein Aliases: dJ3J17.2; Glycine- and tyrosine-rich RNA-binding protein; GRY-RBP; Heterogeneous nuclear ribonucleoprotein Q; hnRNP Q; hnRNP-Q; HNRPQ1; NS1-associated protein 1; NSAP1; pp68; RP1-3J17.2; synaptotagmin binding, cytoplasmic RNA interacting protein; Synaptotagmin-binding, cytoplasmic RNA-interacting protein
Gene Aliases: GRY-RBP; GRYRBP; hnRNP-Q; HNRNPQ; HNRPQ; HNRPQ1; NSAP1; PP68; SYNCRIP
UniProt ID: (Human) O60506
Entrez Gene ID: (Human) 10492
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.