Abnova
Manufacturer Code:H00010492K
Catalog # 89-210-752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SYNCRIP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SYNCRIP peptide using ARM Technology.
For Research Use Only
Protein Aliases: Glycine- and tyrosine-rich RNA-binding protein; GRY-RBP; GRY-RBP HNRPQ1 NSAP1 RP1-3J17.2 dJ3J17.2 hnRNP-Q pp68; Heterogeneous nuclear ribonucleoprotein Q; hnRNP Q; NS1-associated protein 1; synaptotagmin binding, cytoplasmic RNA interacting protein; Synaptotagmin-binding, cytoplasmic RNA-interacting protein
Gene Aliases: GRY-RBP; GRYRBP; hnRNP-Q; HNRNPQ; HNRPQ; HNRPQ1; NSAP1; PP68; SYNCRIP
UniProt ID: (Human) O60506
Entrez Gene ID: (Human) 10492
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.