OriGene
Manufacturer Code:TA806547
Catalog # 50-168-1154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D6 |
Immunogen | Full length human recombit protein of human SYCP3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: COR1 RPRGL4 SCP3 SPGF4; SCP-3; Synaptonemal complex protein 3
Gene Aliases: COR1; RPRGL4; SCP3; SPGF4; SYCP3
UniProt ID: (Human) Q8IZU3
Entrez Gene ID: (Human) 50511
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.