Abnova
Manufacturer Code:PAB12015
Catalog # 89-115-525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human SYCP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of SYCP1.
For Research Use Only
Protein Aliases: Cancer/testis antigen 8; CT8; HOM-TES-14; MGC104417; SCP-1; SCP1; Synaptonemal complex protein 1; testicular tissue protein Li 191
Gene Aliases: CT8; HOM-TES-14; SCP-1; SCP1; SYCP1
UniProt ID: (Human) Q15431
Entrez Gene ID: (Human) 6847
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.