Proteintech
Manufacturer Code:110631AP150UL
Catalog # 50-172-6843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SYCE1 Fusion Protein Ag1527 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C10orf94 Cancer/testis antigen 76 CT76 SYCE1; Cancer/testis antigen 76; CT76; Synaptonemal complex central element protein 1; telomeric protein
Gene Aliases: 4933406J07Rik; C10orf94; CT76; POF12; SPGF15; SYCE1
UniProt ID: (Human) Q8N0S2, (Mouse) Q9D495, (Rat) Q5XHZ2
Entrez Gene ID: (Human) 93426, (Mouse) 74075, (Rat) 502372
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.