Abnova
Manufacturer Code:PAB15585
Catalog # 89-117-659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to full length fission yeast SWI6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against full-length recombinant SWI6.
For Research Use Only
Protein Aliases: Cell-cycle box factor subunit SWI6; MBF subunit P90; PSL8; Regulatory protein SWI6; SDS11; Trans-acting activator of HO endonuclease gene; transcriptional regulator SWI6
Gene Aliases: L9470.8; PSL8; SDS11; SWI6; YLR182W
UniProt ID: (Baker's yeast) P09959
Entrez Gene ID: (Baker's yeast) 850879
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.