Abnova
Manufacturer Code:PAB11378
Catalog # 89-115-055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 314-328 of <i>Schizosaccharomyces pombe</i> swi6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of swi6.
For Research Use Only
Protein Aliases: Chromatin-associated protein swi6; SPAC824.10c
Gene Aliases: SPAC664.01c; SPAC824.10c; swi6
UniProt ID: (Fission yeast) P40381
Entrez Gene ID: (Fission yeast) 2541633
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.