Abnova
Manufacturer Code:H00006604M02
Catalog # 89-000-968
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5F12 |
Immunogen | SMARCD3 (NP_001003801 385 a.a. ~ 483 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant SMARCD3.
For Research Use Only
Protein Aliases: 60 kDa BRG-1/Brm-associated factor subunit C; BAF60C; BRG1-associated factor 60C; chromatin remodeling complex BAF60C subunit; CRACD3; mammalian chromatin remodeling complex BRG1-associated factor 60C; MGC111010; Rsc6p; SWI/SNF complex 60 kDa subunit C; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3, isoform 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 3; Swp73-like protein
Gene Aliases: BAF60C; CRACD3; Rsc6p; SMARCD3
UniProt ID: (Human) Q6STE5
Entrez Gene ID: (Human) 6604
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.