Abnova
Manufacturer Code:H00008467B01
Catalog # 89-006-567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SMARCA5 (AAH23144.1 1 a.a. ~ 1052 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human SMARCA5 protein.
For Research Use Only
Protein Aliases: hISWI; hSNF2H; ISWI; SNF2H; Sucrose nonfermenting protein 2 homolog; sucrose nonfermenting-like 5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 5; WCRF135
Gene Aliases: hISWI; hSNF2H; ISWI; SMARCA5; SNF2H; WCRF135
UniProt ID: (Human) O60264
Entrez Gene ID: (Human) 8467
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.