Invitrogen
Manufacturer Code:PA571964
Catalog # PIPA571964
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 241-269 amino acids from the Central region of human SVOPL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Putative transporter SVOPL; Putative transporter SVOPL SV2-related protein-like SVOP-like protein SVOPL; SV2 related protein homolog-like; SV2-related protein-like; SVOP-like; SVOP-like protein
Gene Aliases: SVOPL
UniProt ID: (Human) Q8N434
Entrez Gene ID: (Human) 136306
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.