Invitrogen
Manufacturer Code:PA569936
Catalog # PIPA569936
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human SVOPL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Putative transporter SVOPL; similar to RIKEN cDNA 9430071P14 gene; SV2 related protein homolog-like; SV2-related protein-like; SVOP-like; SVOP-like protein
Gene Aliases: 9430071P14Rik; si:dkeyp-27e10.2; SVOPL; zgc:101741
UniProt ID: (Human) Q8N434, (Mouse) Q6PDF3, (Zebrafish) Q1LVS8
Entrez Gene ID: (Rat) 500085, (Horse) 100066769, (Human) 136306, (Dog) 482780, (Bovine) 518832, (Rabbit) 100348039, (Mouse) 320590, (Guinea pig) 100721084, (Zebrafish) 492766
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.