Invitrogen
Manufacturer Code:PA5113126
Catalog # PIPA5113126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Putative transporter SVOPL protein (73-129AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9430071P14Rik BOS_4831 Putative transporter SVOPL RGD1566053 si:dkeyp-27e10.2 similar to RIKEN cDNA 9430071P14 gene SV2 related protein homolog (rat)-like SV2 related protein homolog-like SV2-related protein-like SVOP like svopl SVOP-like SVOP-like protein zgc:101741; Putative transporter SVOPL; similar to RIKEN cDNA 9430071P14 gene; SV2 related protein homolog-like; SV2-related protein-like; SVOP-like; SVOP-like protein
Gene Aliases: 9430071P14Rik; SVOPL
UniProt ID: (Human) Q8N434, (Mouse) Q6PDF3
Entrez Gene ID: (Rat) 500085, (Human) 136306, (Mouse) 320590
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.