Abnova
Manufacturer Code:PAB21004
Catalog # 89-122-499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SVOPL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SVOPL.
For Research Use Only
Protein Aliases: MGC46715; Putative transporter SVOPL; SV2 related protein homolog-like; SV2-related protein-like; SVOP-like; SVOP-like protein
Gene Aliases: SVOPL
UniProt ID: (Human) Q8N434
Entrez Gene ID: (Human) 136306
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.